Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Custom Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Who We Serve
Biotech
Biopharma
CDMO
Lab Diagnostics
Industrial and Applied Sciences
Promotions
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Order Status
Quick Order
Promos
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other BANP SIRNA Assays ›
Gene Symbol:
BANP
Gene Name:
BTG3 associated nuclear protein
Gene Aliases:
BEND1, SMAR1, SMARBP1
Chromosome Location:
Chr.16: 87949216 - 88077318 on Build GRCh38
UniGene ID:
Hs.461705
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
44360
Purification
Standard
HPLC
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: BANP
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001173539.1
NP_001167010.1
3
241
NM_001173540.1
NP_001167011.1
3
259
NM_001173541.1
NP_001167012.1
3
241
NM_001173542.1
NP_001167013.1
3
280
NM_001173543.1
NP_001167014.1
3
310
NM_017869.3
NP_060339.2
3
241
NM_079837.2
NP_524576.2
3
241
XM_005256008.3
XP_005256065.1
3
257
XM_005256009.3
XP_005256066.1
3
239
XM_011523170.2
XP_011521472.1
3
257
XM_011523172.2
XP_011521474.1
4
502
XM_011523174.2
XP_011521476.1
3
1028
XM_011523175.2
XP_011521477.1
3
659
XM_011523176.2
XP_011521478.1
3
721
XM_011523177.2
XP_011521479.1
3
1229
XM_011523178.2
XP_011521480.1
3
257
XM_011523179.2
XP_011521481.1
3
239
XM_011523180.2
XP_011521482.1
4
321
XM_011523181.2
XP_011521483.1
3
257
XM_011523182.1
XP_011521484.1
3
257
XM_011523183.2
XP_011521485.1
3
234
XM_011523184.2
XP_011521486.1
3
190
XM_011523186.2
XP_011521488.1
1
19
XM_017023378.1
XP_016878867.1
4
361
XM_017023379.1
XP_016878868.1
3
641
XM_017023380.1
XP_016878869.1
3
241
XM_017023381.1
XP_016878870.1
3
259
XM_017023383.1
XP_016878872.1
3
206
GenBank mRNA
AK001039.1
-
3
241
AK094158.1
-
3
311
AK125232.1
-
3
239
AK293472.1
-
3
203
AK297945.1
-
3
213
AK315089.1
-
3
239
AK315535.1
-
3
280
BC009424.1
-
3
186
3530821
DA632793.1
-
3
241
KU178664.1
-
2
89
KU178665.1
-
2
89
KU178666.1
-
2
89
KU178667.1
-
2
89
KU178668.1
-
2
89
KU178669.1
-
2
89
KU178670.1
-
2
111
Back To Top
More Information
Gene Ontology Categories:
Function(s)
Process(es)
transcription, DNA-templated
cell cycle
multicellular organism development
covalent chromatin modification
protein localization to nucleus
negative regulation of protein catabolic process
positive regulation of transcription, DNA-templated
regulation of signal transduction by p53 class mediator
p53 binding
DNA binding
protein binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-green-659f68c6f4-wf7gw:80/100.66.73.223:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0