siRNA Details
Gene Symbol: ZMAT3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_022470.3 | NP_071915.1 | 2, 3 | 597 | ||
NM_152240.2 | NP_689426.1 | 3, 4 | 718 | |||
XM_005247706.2 | XP_005247763.1 | 2, 3 | 1008 | |||
XM_011513073.1 | XP_011511375.1 | 3, 4 | 1129 | |||
GenBank mRNA | AF355465.1 | - | 2, 3 | 406 | ||
AK022358.1 | - | 2, 3 | 524 | |||
AK122768.1 | - | 3, 4 | 607 | |||
AY037945.1 | - | 1, 2 | 296 | |||
BC002896.2 | - | 2, 3 | 525 | 3943641 |

More Information
Panther Classification:
Molecular Function -




