Search
Search

GLT8D1| Gene Symbol: GLT8D1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001010983.2 | NP_001010983.1 | 8, 9 | 1555 | ||
| NM_001278280.1 | NP_001265209.1 | 8, 9 | 1551 | |||
| NM_001278281.1 | NP_001265210.1 | 8, 9 | 1299 | |||
| NM_018446.3 | NP_060916.1 | 7, 8 | 1326 | |||
| NM_152932.2 | NP_690909.1 | 7, 8 | 884 | |||
| XM_017006857.1 | XP_016862346.1 | 9, 10 | 1714 | |||
| XM_017006858.1 | XP_016862347.1 | 8, 9 | 1485 | |||
| XM_017006859.1 | XP_016862348.1 | 7, 8 | 1034 | |||
| GenBank mRNA | AF157318.1 | - | 7, 8 | 771 | ||
| AF175227.1 | - | 8, 9 | 1038 | |||
| AF176777.1 | - | 8, 9 | 1041 | |||
| AJ271710.1 | - | 6, 7 | 653 | |||
| AK027517.1 | - | 7, 8 | 792 | |||
| AY358579.1 | - | 7, 8 | 805 | |||
| BC119670.1 | - | 7, 8 | 770 | 40116196 | ||
| BC119671.1 | - | 7, 8 | 770 | 40116197 | ||
| CR749821.1 | - | 8, 9 | 1008 | |||