Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
CDMO and Clinical Trial Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Instrument Management
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other PTPRD SIRNA Assays ›
Gene Symbol:
PTPRD
Gene Name:
protein tyrosine phosphatase, receptor type D
Gene Aliases:
HPTP, HPTPD, HPTPDELTA, PTPD, RPTPDELTA
Chromosome Location:
Chr.9: 8314246 - 10612840 on Build GRCh38
UniGene ID:
Hs.446083
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
4453
Purification
HPLC
Standard
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: PTPRD
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001040712.2
NP_001035802.1
6
709
NM_001171025.1
NP_001164496.1
6
700
NM_002839.3
NP_002830.1
18
1358
NM_130391.3
NP_569075.2
7
718
NM_130392.3
NP_569076.2
7
718
NM_130393.3
NP_569077.2
6
700
XM_006716817.3
XP_006716880.1
17
1656
XM_006716823.2
XP_006716886.1
17
1650
XM_006716825.3
XP_006716888.1
17
1652
XM_006716827.3
XP_006716890.1
15
1622
XM_006716832.3
XP_006716895.1
16
1626
XM_006716833.3
XP_006716896.1
17
1646
XM_006716834.3
XP_006716897.1
15
1620
XM_006716835.3
XP_006716898.1
16
1637
XM_006716837.3
XP_006716900.1
16
1626
XM_006716838.3
XP_006716901.1
15
1619
XM_006716839.3
XP_006716902.1
15
1617
XM_011517992.2
XP_011516294.1
17
1649
XM_017014958.1
XP_016870447.1
17
1655
XM_017014959.1
XP_016870448.1
18
1727
XM_017014960.1
XP_016870449.1
16
1616
XM_017014961.1
XP_016870450.1
16
1600
XM_017014962.1
XP_016870451.1
8
788
XM_017014963.1
XP_016870452.1
17
1655
XM_017014964.1
XP_016870453.1
17
1654
XM_017014965.1
XP_016870454.1
17
1657
XM_017014966.1
XP_016870455.1
16
1646
XM_017014967.1
XP_016870456.1
17
1653
XM_017014968.1
XP_016870457.1
17
1652
XM_017014969.1
XP_016870458.1
16
1637
XM_017014970.1
XP_016870459.1
17
1655
XM_017014971.1
XP_016870460.1
15
1626
XM_017014972.1
XP_016870461.1
17
1655
XM_017014973.1
XP_016870462.1
16
1634
XM_017014974.1
XP_016870463.1
17
1655
XM_017014975.1
XP_016870464.1
15
1623
XM_017014976.1
XP_016870465.1
17
1652
XM_017014977.1
XP_016870466.1
17
1653
XM_017014978.1
XP_016870467.1
17
1652
XM_017014979.1
XP_016870468.1
17
1654
XM_017014980.1
XP_016870469.1
17
1649
XM_017014981.1
XP_016870470.1
15
1617
XM_017014982.1
XP_016870471.1
17
1651
XM_017014983.1
XP_016870472.1
17
1651
XM_017014984.1
XP_016870473.1
16
1642
XM_017014985.1
XP_016870474.1
17
1648
XM_017014986.1
XP_016870475.1
17
1647
XM_017014987.1
XP_016870476.1
16
1629
XM_017014988.1
XP_016870477.1
16
1629
XM_017014989.1
XP_016870478.1
17
1647
XM_017014990.1
XP_016870479.1
17
1646
XM_017014991.1
XP_016870480.1
17
1646
XM_017014992.1
XP_016870481.1
16
1637
XM_017014993.1
XP_016870482.1
17
1644
XM_017014994.1
XP_016870483.1
17
1644
XM_017014995.1
XP_016870484.1
16
1641
GenBank mRNA
AB208869.1
-
5
535
AB211400.1
-
17
1326
BC106713.1
-
6
648
40027581
BC106714.1
-
7
666
40027582
BC106715.2
-
7
666
40027584
BC106716.1
-
6
648
40027585
L38929.1
-
8
768
Back To Top
More Information
Gene Ontology Categories:
Function(s)
Process(es)
protein dephosphorylation
phosphate-containing compound metabolic process
heterophilic cell-cell adhesion via plasma membrane cell adhesion molecules
transmembrane receptor protein tyrosine phosphatase signaling pathway
chemical synaptic transmission
neuron differentiation
peptidyl-tyrosine dephosphorylation
positive regulation of dendrite morphogenesis
presynaptic membrane assembly
transmembrane receptor protein tyrosine phosphatase activity
receptor binding
protein binding
cell adhesion molecule binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-65b4bdbffd-d6q6k:80/10.239.161.116:80.
git-commit: b16971a8df2afcd8df684aafbc9bf1e9f45e42d4
git-url: git@github.com:thermofisher/magellan-core.git
git-branch: origin/release/2.6.0-2024.04.15-1.0