Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture and Transfection
    • Chemicals
    • Chromatography
    • Electron Microscopes
    • Lab Plasticware and Supplies
    • Lab Centrifuges
    • Lab Solutions
    • Mass Spectrometers
    • Next Generation Sequencers
    • See all product categories
  • Applications
    • Brands
    • Industrial and Applied Sciences
    • Food and Beverage
    • Forensics
    • Lab Solutions
    • Life Sciences
    • Pharma and Biopharma
    • Biotechnology
    • Clinical and Diagnostics
    • Digital Solutions
    • See all applications
  • Services
    • Lab Instrument and Equipment Services
    • Enterprise Services
    • Custom Services
    • Training Services
    • Financial and Leasing Services
    • Enterprise Level Lab Informatics
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Cell Biology Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • Contact Us
    • Product Documentation
    • Knowledge Base and Product FAQs
    • Learning Centers
    • Supply Center
    • eProcurement Solutions
    • Lab Instrument and Equipment Support
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Special Offers
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other FGF1 SIRNA Assays ›
          Gene Symbol:
          FGF1
          Gene Name:
          fibroblast growth factor 1
          Gene Aliases:
          AFGF, ECGF, ECGF-beta, ECGFA, ECGFB, FGF-1, FGF-alpha, FGFA, GLIO703, HBGF-1, HBGF1
          Chromosome Location:
          Chr.5: 142592178 - 142698070 on Build GRCh38
          UniGene ID:
          Hs.483635
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID 44677

          Purification
          Size
          Availability Inventoried
          Catalog # AM16708
          Price 4,126.00
          Your Price
          Online offer:
          4126.0
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: FGF1
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_000800.4 NP_000791.1 4 1565
          NM_001144892.2 NP_001138364.1 4 1085
          NM_001144934.1 NP_001138406.1 5 1278
          NM_001144935.1 NP_001138407.1 4 1184
          NM_001257205.1 NP_001244134.1 4 1475
          NM_001257206.1 NP_001244135.1 4 1472
          NM_001257207.1 NP_001244136.1 5 1218
          NM_001257208.1 NP_001244137.1 4 1216
          NM_001257209.1 NP_001244138.1 4 1231
          NM_001257210.1 NP_001244139.1 4 1215
          NM_001257211.1 NP_001244140.1 4 1213
          NM_001257212.1 NP_001244141.1 4 1082
          NM_033136.3 NP_149127.1 3 1461
          NM_033137.2 NP_149128.1 2 919
          XM_005268390.4 XP_005268447.1 4 1155
          XM_011537607.2 XP_011535909.1 5 1275
          GenBank mRNA X51943.1 - 3 1026

          Back To Top

          More Information


          Panther Classification:

          Molecular Function -

          growth factor intercellular signal molecule

          Gene Ontology Categories:

          Function(s) Process(es)

          MAPK cascade
          activation of MAPK activity
          angiogenesis
          organ induction
          signal transduction
          multicellular organism development
          cell proliferation
          positive regulation of cell proliferation
          fibroblast growth factor receptor signaling pathway
          anatomical structure morphogenesis
          positive regulation of endothelial cell migration
          regulation of phosphatidylinositol 3-kinase signaling
          peptidyl-tyrosine phosphorylation
          cell differentiation
          lung development
          positive regulation of cell migration
          activation of protein kinase B activity
          cellular response to heat
          phosphatidylinositol-3-phosphate biosynthetic process
          positive regulation of MAP kinase activity
          positive regulation of GTPase activity
          positive regulation of cholesterol biosynthetic process
          positive regulation of angiogenesis
          positive regulation of transcription from RNA polymerase II promoter
          phosphatidylinositol phosphorylation
          phosphatidylinositol-mediated signaling
          positive regulation of epithelial cell proliferation
          positive regulation of cell division
          branch elongation involved in ureteric bud branching
          positive regulation of ERK1 and ERK2 cascade
          mesonephric epithelium development
          regulation of endothelial tube morphogenesis
          positive regulation of intracellular signal transduction
          positive regulation of sprouting angiogenesis
          regulation of endothelial cell chemotaxis to fibroblast growth factor
          protein tyrosine kinase activity
          Ras guanyl-nucleotide exchange factor activity
          fibroblast growth factor receptor binding
          integrin binding
          protein binding
          growth factor activity
          heparin binding
          1-phosphatidylinositol-3-kinase activity
          Hsp70 protein binding
          S100 protein binding
          phosphatidylinositol-4,5-bisphosphate 3-kinase activity

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Quick Order
          • eProcurement
          • Supply Center
          • Order Status
          • Chemicals
          • India Mobile App
          • Government eMarketplace
          Support Plus Icon Minus Icon
          • Order Support
          • Training
          • Contact Us
          • Report a Site Issue
          • Instrument Management
          Resources Plus Icon Minus Icon
          • Product Selection Guides
          • Mobile & Desktop Apps
          • Webinars
          • Blog 
          • Social Media
          • New Products
          • Promotions
          • Shared Lists
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          India flag icon
          India

          Your items have has been added!


          Host server : magellan-search-blue-55ff9ddd88-lffdw:80/100.66.79.31:80.
          git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.42.0-2026.01.03-1.0