Hamburger Menu Button
Thermo Fisher Scientific Logo
登入
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Tubes
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Clinical Genomics
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO 和 CRO 服務
    • CDMO 服務
    • 臨床試驗 CRO 服務
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • Register for an Account
    • How to Order
    • Instrument Support
    • Technical Support Centers
    • Learning Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • 快速訂購
          • 登入
            登入
            Don't have an account ? Create Account
            • 帳戶
            • 訂單
            • 訂製產品與專案
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other CHRFAM7A SIRNA Assays ›
          Gene Symbol:
          CHRFAM7A CHRNA7
          Gene Name:
          CHRNA7 (exons 5-10) and FAM7A (exons A-E) fusion
          cholinergic receptor nicotinic alpha 7 subunit
          Gene Aliases:
          CHRNA7, CHRNA7-DR1, D-10
          CHRNA7-2, NACHRA7
          Chromosome Location:
          Chr.15: 30357770 - 30393849 on Build GRCh38
          Chromosome Location:
          Chr.15: 32030462 - 32172521 on Build GRCh38
          UniGene ID:
          Hs.510853
          Hs.511772
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          -
          siRNA ID 44727

          Purification
          Size
          Availability Inventoried
          Catalog # AM16708
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: CHRFAM7A
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_139320.1 NP_647536.1 10 2428
          NM_148911.1 NP_683709.1 9 2364
          XM_005254750.2 XP_005254807.1 11 2597
          XM_011522153.1 XP_011520455.1 11 2122
          XM_011522155.2 XP_011520457.1 9 2313
          GenBank mRNA AK292984.1 - 10 2193
          BQ720423.1 - 1 319
          Gene Symbol: CHRNA7
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_000746.5 NP_000737.1 10 2242
          NM_001190455.2 NP_001177384.1 10 2329
          NR_046324.1 8 2052
          XM_011521176.2 XP_011519478.2 9 2097
          XM_011521177.2 XP_011519479.1 7 2182
          XM_017021882.1 XP_016877371.1 11 2307
          XM_017021883.1 XP_016877372.1 11 2304
          XM_017021884.1 XP_016877373.1 9 2153

          Back To Top

          More Information


          Panther Classification:

          Molecular Function -

          ligand-gated ion channel

          Gene Ontology Categories:

          Function(s) Process(es)

          ion transmembrane transport
          activation of MAPK activity
          response to hypoxia
          ion transport
          calcium ion transport
          cellular calcium ion homeostasis
          signal transduction
          synaptic transmission, cholinergic
          memory
          positive regulation of cell proliferation
          negative regulation of tumor necrosis factor production
          response to nicotine
          positive regulation of angiogenesis
          cognition
          cation transmembrane transport
          extracellular ligand-gated ion channel activity
          beta-amyloid binding
          acetylcholine-activated cation-selective channel activity
          ligand-gated ion channel activity
          acetylcholine receptor activity
          toxic substance binding
          chloride channel regulator activity
          acetylcholine-gated cation channel activity
          acetylcholine binding
          protein homodimerization activity

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Hong Kong flag icon
          Hong Kong

          Your items have has been added!


          Host server : magellan-search-blue-55ff9ddd88-nq57x:80/100.66.75.150:80.
          git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.42.0-2026.01.03-1.0