Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Western Blot Products
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Food and Beverage
    • Lab Solutions
    • Pharma and Biopharma
    • Real-Time PCR
    • Semiconductor Analysis
    • Clinical and Diagnostics
    • Digital Solutions
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • See all services
  • Help and Support
    • Order Help
    • Digital Solutions
    • Product Support
    • Technical Information
    • Training and Education
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Special Offers
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other INS SIRNA Assays ›
          Gene Symbol:
          INS INS-IGF2
          Gene Name:
          insulin
          INS-IGF2 readthrough
          Gene Aliases:
          IDDM, IDDM1, IDDM2, ILPR, IRDN, MODY10
          INSIGF
          Chromosome Location:
          Chr.11: 2159779 - 2161209 on Build GRCh38
          Chromosome Location:
          Chr.11: 2129112 - 2161209 on Build GRCh38
          UniGene ID:
          Hs.272259
          Hs.272259
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          -
          siRNA ID 44761

          Purification
          Size
          Availability Inventoried
          Catalog # AM16708
          Price 4,126.00
          Your Price
          Online offer:
          4126.0
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: INS
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_000207.2 NP_000198.1 2 137
          NM_001185097.1 NP_001172026.1 2 163
          NM_001185098.1 NP_001172027.1 1 316
          NM_001291897.1 NP_001278826.1 2 197
          GenBank mRNA AY899304.1 - 2 133
          BC005255.1 - 2 137 3950204
          BM510347.1 - 2 158
          BM510748.1 - 2 141
          BP322143.1 - 1 316
          BT006808.1 - 1 78
          DQ778082.1 - 1 78
          JF909299.1 - 1 31
          JQ951950.1 - 2 122
          X70508.1 - 2 122
          Gene Symbol: INS-IGF2
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001042376.2 NP_001035835.1 2 137
          NR_003512.3 2 137
          GenBank mRNA DQ104204.1 - 2 137
          DQ104205.1 - 2 137

          Back To Top

          More Information


          Panther Classification:

          Molecular Function -

          growth factor intercellular signal molecule

          Gene Ontology Categories:

          Function(s) Process(es)

          MAPK cascade
          negative regulation of acute inflammatory response
          glucose metabolic process
          regulation of transcription, DNA-templated
          regulation of cellular amino acid metabolic process
          ER to Golgi vesicle-mediated transport
          acute-phase response
          G-protein coupled receptor signaling pathway
          cell-cell signaling
          positive regulation of cell proliferation
          insulin receptor signaling pathway
          positive regulation of gene expression
          positive regulation of phosphatidylinositol 3-kinase signaling
          glucose transport
          regulation of transmembrane transporter activity
          positive regulation of cell growth
          positive regulation of cell migration
          positive regulation of protein autophosphorylation
          activation of protein kinase B activity
          positive regulation of cellular protein metabolic process
          negative regulation of protein oligomerization
          regulation of protein localization
          negative regulation of NAD(P)H oxidase activity
          wound healing
          negative regulation of protein catabolic process
          glucose homeostasis
          positive regulation of MAPK cascade
          cellular protein metabolic process
          positive regulation of nitric oxide biosynthetic process
          positive regulation of cell differentiation
          negative regulation of gluconeogenesis
          positive regulation of glycogen biosynthetic process
          positive regulation of DNA replication
          negative regulation of glycogen catabolic process
          positive regulation of glycolytic process
          positive regulation of mitotic nuclear division
          negative regulation of proteolysis
          negative regulation of vasodilation
          positive regulation of vasodilation
          negative regulation of fatty acid metabolic process
          positive regulation of glucose import
          positive regulation of insulin receptor signaling pathway
          alpha-beta T cell activation
          positive regulation of lipid biosynthetic process
          regulation of protein secretion
          negative regulation of protein secretion
          positive regulation of cytokine secretion
          positive regulation of peptidyl-tyrosine phosphorylation
          negative regulation of lipid catabolic process
          positive regulation of nitric-oxide synthase activity
          positive regulation of NF-kappaB transcription factor activity
          positive regulation of protein kinase B signaling
          fatty acid homeostasis
          negative regulation of respiratory burst involved in inflammatory response
          positive regulation of respiratory burst
          positive regulation of peptide hormone secretion
          positive regulation of brown fat cell differentiation
          positive regulation of protein localization to nucleus
          negative regulation of oxidative stress-induced intrinsic apoptotic signaling pathway
          negative regulation of feeding behavior
          protease binding
          insulin receptor binding
          insulin-like growth factor receptor binding
          hormone activity
          protein binding
          identical protein binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Taiwan flag icon
          Taiwan

          Your items have has been added!


          Host server : magellan-search-blue-55ff9ddd88-8jdkq:80/100.66.75.107:80.
          git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.42.0-2026.01.03-1.0