Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture and Transfection
Chemicals
Chromatography
Electron Microscopes
Lab Plasticware and Supplies
Lab Centrifuges
Lab Solutions
Mass Spectrometers
Next Generation Sequencers
See all product categories
Applications
Brands
Industrial and Applied Sciences
Food and Beverage
Forensics
Lab Solutions
Life Sciences
Pharma and Biopharma
Biotechnology
Clinical and Diagnostics
Digital Solutions
See all applications
Services
Lab Instrument and Equipment Services
Enterprise Services
Custom Services
Training Services
Financial and Leasing Services
Enterprise Level Lab Informatics
Partnering and Licensing Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Cell Biology Services
Food Safety Inspection Services
See all services
Help and Support
Contact Us
Product Documentation
Knowledge Base and Product FAQs
Learning Centers
Supply Center
eProcurement Solutions
Lab Instrument and Equipment Support
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Special Offers
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Custom Products & Projects
Instrument Management
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other PPOX SIRNA Assays ›
Gene Symbol:
PPOX
Gene Name:
protoporphyrinogen oxidase
Gene Aliases:
PPO, V290M, VP
Chromosome Location:
Chr.1: 161165728 - 161178277 on Build GRCh38
UniGene ID:
Hs.517373
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
44763
Purification
Standard
HPLC
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
286.00
Your Price
Online offer:
277.65
277.65
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: PPOX
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_000309.3
NP_000300.1
3
379
NM_001122764.1
NP_001116236.1
3
440
XM_005245291.3
XP_005245348.2
3
418
XM_005245295.3
XP_005245352.2
3
363
XM_006711402.2
XP_006711465.2
3
370
XM_006711403.2
XP_006711466.2
3
374
XM_006711404.3
XP_006711467.2
3
307
XM_006711406.2
XP_006711469.2
3
276
XM_011509663.2
XP_011507965.1
3
490
XM_011509664.1
XP_011507966.1
3
276
XM_011509665.2
XP_011507967.1
3
488
XM_011509666.2
XP_011507968.1
3
495
XM_011509667.2
XP_011507969.1
3
431
XM_011509668.2
XP_011507970.1
3
231
XM_011509669.1
XP_011507971.1
3
244
XM_011509670.2
XP_011507972.1
3
493
XM_011509671.1
XP_011507973.1
3
276
XM_011509672.2
XP_011507974.1
3
484
XM_011509673.2
XP_011507975.1
3
491
XM_011509674.2
XP_011507976.1
3
496
XM_011509675.2
XP_011507977.1
3
426
XM_011509676.1
XP_011507978.1
3
435
XM_011509677.1
XP_011507979.1
3
368
XM_011509678.1
XP_011507980.1
3
432
XM_011509679.1
XP_011507981.1
3
240
XM_011509681.1
XP_011507983.1
3
290
XM_011509682.1
XP_011507984.1
3
363
XM_017001559.1
XP_016857048.1
3
487
XM_017001560.1
XP_016857049.1
3
418
XM_017001561.1
XP_016857050.1
3
371
XM_017001562.1
XP_016857051.1
3
345
XM_017001563.1
XP_016857052.1
3
430
XM_017001565.1
XP_016857054.1
3
345
XM_017001566.1
XP_016857055.1
3
422
XM_017001567.1
XP_016857056.1
3
363
XM_017001568.1
XP_016857057.1
3
422
XM_017001569.1
XP_016857058.1
3
290
XM_017001570.1
XP_016857059.1
3
403
XM_017001571.1
XP_016857060.1
3
508
GenBank mRNA
AK094855.1
-
3
294
AK298912.1
-
3
320
AK302298.1
-
3
349
BC002357.2
-
3
360
2821983
BX384814.2
-
3
439
D38537.1
-
3
427
DC329287.1
-
3
451
U26446.1
-
3
341
Back To Top
More Information
Panther Classification:
Molecular Function -
oxidase
Gene Ontology Categories:
Function(s)
Process(es)
porphyrin-containing compound biosynthetic process
protoporphyrinogen IX biosynthetic process
heme biosynthetic process
response to drug
oxidation-reduction process
oxygen-dependent protoporphyrinogen oxidase activity
flavin adenine dinucleotide binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-blue-55ff9ddd88-ctdjb:80/100.66.76.55:80.
git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.42.0-2026.01.03-1.0