siRNA Details
Gene Symbol: AQP2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000486.5 | NP_000477.1 | 4 | 1218 | ||
GenBank mRNA | AK225940.1 | - | 5 | 855 | ||
BC042496.1 | - | 4 | 1213 | 5186409 |

Gene Symbol: AQP2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000486.5 | NP_000477.1 | 4 | 1218 | ||
GenBank mRNA | AK225940.1 | - | 5 | 855 | ||
BC042496.1 | - | 4 | 1213 | 5186409 |