siRNA Details
Gene Symbol: POU3F1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_002699.3 | NP_002690.3 | 1 | 1512 | ||
GenBank mRNA | L26494.1 | - | 1 | 1504 |

Gene Symbol: POU3F1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_002699.3 | NP_002690.3 | 1 | 1512 | ||
GenBank mRNA | L26494.1 | - | 1 | 1504 |