Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FCRL2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001159488.1 | NP_001152960.1 | 7 | 1248 | ||
NM_030764.3 | NP_110391.2 | 7 | 1248 | |||
NR_125358.1 | 4 | 396 | ||||
XM_011509974.2 | XP_011508276.1 | 7 | 1583 | |||
XM_011509975.2 | XP_011508277.1 | 7 | 1585 | |||
XM_011509976.2 | XP_011508278.1 | 7 | 1584 | |||
XM_017002316.1 | XP_016857805.1 | 7 | 1583 | |||
XM_017002317.1 | XP_016857806.1 | 7 | 1386 | |||
XM_017002318.1 | XP_016857807.1 | 6 | 1306 | |||
XM_017002319.1 | XP_016857808.1 | 6 | 1298 | |||
GenBank mRNA | AF319438.1 | - | 2 | 733 | ||
AF319439.1 | - | 2 | 571 | |||
AF319440.1 | - | 1 | 980 | |||
AF390037.1 | - | 2 | 571 | |||
AF459633.1 | - | 7 | 1236 | |||
AK301113.1 | - | 7 | 1235 | |||
AL833194.1 | - | 2 | 3107 | |||
AY043465.1 | - | 7 | 1235 | |||
AY358130.1 | - | 7 | 1238 | |||
BC069185.1 | - | 4 | 400 | |||
BC129836.1 | - | 2 | 573 | 40115446 |