Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: OR2J2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_030905.2 | NP_112167.2 | 1 | 179 | ||
GenBank mRNA | BC069121.1 | - | 1 | 77 | 7216975 | |
BC108897.1 | - | 1 | 77 | 40003955 | ||
BC108898.1 | - | 1 | 77 | 40003956 | ||
BC108899.1 | - | 1 | 77 | 40003957 | ||
BU171389.1 | - | 1 | 181 |