Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RBBP6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_006910.4 | NP_008841.2 | 1 | 1155 | ||
NM_018703.3 | NP_061173.1 | 1 | 1155 | |||
NM_032626.5 | NP_116015.2 | 1 | 1155 | |||
GenBank mRNA | AB112074.1 | - | 1 | 159 | ||
AB112075.1 | - | 1 | 159 | |||
AF063596.1 | - | 1 | 390 | |||
BC029352.1 | - | 1 | 246 | |||
BC063524.1 | - | 1 | 222 | |||
BC101139.2 | - | 1 | 277 | |||
BC101140.2 | - | 1 | 277 | |||
BC101141.2 | - | 1 | 277 | |||
BC101142.1 | - | 1 | 277 | |||
BC114353.1 | - | 1 | 277 | |||
BC114354.1 | - | 1 | 277 | |||
BC118667.1 | - | 1 | 277 | |||
BC139830.1 | - | 1 | 277 | |||
BM314442.1 | - | 1 | 525 | |||
BX648661.1 | - | 1 | 95 |