Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PCGF1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_032673.2 | NP_116062.2 | 2 | 140 | ||
GenBank mRNA | AF087884.1 | - | 1 | 246 | ||
AK125742.1 | - | 2 | 1026 | |||
BC004952.1 | - | 2 | 105 | 3621400 | ||
BU602498.1 | - | 2 | 143 |