Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Enterprise Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other NR4A3 SIRNA Assays ›
          Gene Symbol:
          NR4A3
          Gene Name:
          nuclear receptor subfamily 4 group A member 3
          Gene Aliases:
          CHN, CSMF, MINOR, NOR1, TEC
          Chromosome Location:
          Chr.9: 99821855 - 99866893 on Build GRCh38
          UniGene ID:
          Hs.279522
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID 45758

          Purification
          Size
          Availability Inventoried
          Catalog # AM16708
          Price 286.00
          Your Price
          Online offer: 277.65
          277.65
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: NR4A3
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_006981.3 NP_008912.2 3 784
          NM_173199.2 NP_775291.1 3 784
          NM_173200.2 NP_775292.1 4 856
          XM_017015162.1 XP_016870651.1 2 716
          GenBank mRNA AK226108.1 - 3 766
          D78579.1 - 3 786
          D85241.1 - 1 812
          S81243.1 - 3 671
          X89894.1 - 3 742

          Back To Top

          More Information


          Panther Classification:

          Molecular Function -

          C4 zinc finger nuclear receptor

          Gene Ontology Categories:

          Function(s) Process(es)

          negative regulation of transcription from RNA polymerase II promoter
          mesoderm formation
          transcription initiation from RNA polymerase II promoter
          gastrulation
          axon guidance
          biological_process
          pyruvate oxidation
          positive regulation of cardiac muscle hypertrophy
          positive regulation of glucose transport
          hippocampus development
          intracellular receptor signaling pathway
          adult behavior
          organ regeneration
          positive regulation of mast cell cytokine production
          common myeloid progenitor cell proliferation
          positive regulation of mast cell activation by Fc-epsilon receptor signaling pathway
          response to hydrogen peroxide
          mast cell degranulation
          steroid hormone mediated signaling pathway
          negative regulation of neuron apoptotic process
          cellular response to leptin stimulus
          cellular respiration
          fat cell differentiation
          positive regulation of cell cycle
          positive regulation of transcription from RNA polymerase II promoter
          positive regulation of fatty acid oxidation
          regulation of smooth muscle cell proliferation
          positive regulation of smooth muscle cell proliferation
          semicircular canal morphogenesis
          positive regulation of epithelial cell proliferation
          neuromuscular process controlling balance
          vestibular reflex
          regulation of type B pancreatic cell proliferation
          cellular response to corticotropin-releasing hormone stimulus
          cellular response to catecholamine stimulus
          energy homeostasis
          positive regulation of monocyte aggregation
          negative regulation of hydrogen peroxide-induced neuron death
          positive regulation of leukocyte apoptotic process
          positive regulation of feeding behavior
          regulation of energy homeostasis
          RNA polymerase II core promoter proximal region sequence-specific DNA binding
          core promoter sequence-specific DNA binding
          transcriptional activator activity, RNA polymerase II core promoter proximal region sequence-specific binding
          transcription coactivator binding
          DNA binding
          steroid hormone receptor activity
          thyroid hormone receptor activity
          protein binding
          zinc ion binding
          protein kinase binding
          histone acetyltransferase binding
          glucocorticoid receptor binding
          protein homodimerization activity

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          México flag icon
          México

          Your items have has been added!


          Host server : magellan-search-blue-55ff9ddd88-ctdjb:80/100.66.76.55:80.
          git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.42.0-2026.01.03-1.0