Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ADH1A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000667.3 | NP_000658.1 | 4 | 390 | ||
GenBank mRNA | AK290558.1 | - | 4 | 347 | ||
AK309597.1 | - | 4 | 365 | |||
BC074738.2 | - | 4 | 304 | 30915195 | ||
BC117442.1 | - | 4 | 346 | 40125993 | ||
BC126306.1 | - | 4 | 346 | 8992022 | ||
BT019812.1 | - | 4 | 276 | |||
M12271.1 | - | 4 | 291 | |||
M12963.1 | - | 4 | 348 |