siRNA Details
Gene Symbol: TCF7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001134851.2 | NP_001128323.2 | 9 | 1321 | ||
NM_003202.3 | NP_003193.2 | 9, 10 | 1282 | |||
NM_201632.3 | NP_963963.1 | 8, 9 | 838 | |||
NM_201634.3 | NP_963965.1 | 10 | 934 | |||
NM_213648.3 | NP_998813.1 | 7, 8 | 1072 | |||
NR_033449.1 | 7, 8 | 1073 | ||||
XM_006714680.3 | XP_006714743.1 | 10, 11 | 1383 | |||
XM_006714682.2 | XP_006714745.1 | 11 | 1406 | |||
XM_006714684.2 | XP_006714747.1 | 10 | 1313 | |||
XM_011543606.1 | XP_011541908.1 | 11 | 1796 | |||
XM_011543612.2 | XP_011541914.1 | 7, 8 | 784 | |||
XM_017009789.1 | XP_016865278.1 | 10 | 951 | |||
XM_017009790.1 | XP_016865279.1 | 8, 9 | 1810 | |||
GenBank mRNA | AK057580.1 | - | 7, 8 | 1073 | ||
AK093530.1 | - | 7, 8 | 943 | |||
AK093683.1 | - | 4, 5 | 1172 | |||
AK131428.1 | - | 5, 6 | 1364 | |||
AK310591.1 | - | 7, 8 | 784 | |||
AL832816.1 | - | 9, 10 | 998 | |||
AL834166.1 | - | 8 | 3591 | |||
BC048769.1 | - | 9, 10 | 1282 | 6063270 | ||
BC072023.1 | - | 9, 10 | 1217 | 6292932 | ||
DB127975.1 | - | 1 | 287 | |||
X59870.1 | - | 8, 9 | 820 | |||
X59871.1 | - | 10 | 916 | |||
Z47361.1 | - | 8 | 765 | |||
Z47362.1 | - | 8, 9 | 815 | |||
Z47363.1 | - | 2, 3 | 115 | |||
Z47364.1 | - | 2 | 447 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
