Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CLPP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_006012.2 | NP_006003.1 | 3 | 421 | ||
GenBank mRNA | AK311973.1 | - | 3 | 321 | ||
BC002956.1 | - | 3 | 421 | 3542292 | ||
Z50853.1 | - | 3 | 317 |