Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
CDMO and Clinical Trial Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Instrument Management
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other LILRB3 SIRNA Assays ›
Gene Symbol:
LILRB3
LILRA6
Gene Name:
leukocyte immunoglobulin like receptor B3
leukocyte immunoglobulin like receptor A6
Gene Aliases:
CD85A, HL9, ILT-5, ILT5, LILRA6, LIR-3, LIR3, PIR-B, PIRB
CD85b, ILT-8, ILT5, ILT8, LILRB3, LILRB6
Chromosome Location:
Chr.19: 54216278 - 54223087 on Build GRCh38
Chromosome Location:
Chr.19: 54236592 - 54242768 on Build GRCh38
UniGene ID:
Hs.631592
Hs.688335
Species:
Human
Species Specific ID (Flybase ID):
-
-
siRNA ID
46172
Purification
HPLC
Standard
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: LILRB3
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001081450.2
NP_001074919.2
8
1438
NM_001320960.1
NP_001307889.1
9
1489
NM_006864.3
NP_006855.3
8
1438
NR_135493.1
8
1438
NR_135494.1
8
1352
NR_135495.1
8
1358
NR_135496.1
9
1521
XM_011526381.2
XP_011524683.1
9
1489
XM_011526382.2
XP_011524684.1
9
1489
XM_017026218.1
XP_016881707.1
8
1357
XM_017026219.1
XP_016881708.1
8
1357
XM_017026220.1
XP_016881709.1
8
1357
XM_017026221.1
XP_016881710.1
8
1357
XM_017026222.1
XP_016881711.1
7
1306
XM_017026223.1
XP_016881712.1
7
1306
GenBank mRNA
AF000575.1
-
8
1327
AF009632.1
-
8
1329
AF009633.1
-
8
1329
AF009634.1
-
8
1329
AF009635.1
-
8
1329
AF009636.1
-
8
1329
AF009637.1
-
9
1380
AF009638.1
-
8
1329
AF009639.1
-
8
1329
AF009640.1
-
8
1329
AF009641.1
-
8
1329
AF009642.1
-
8
1329
AF009643.1
-
8
1243
AF009644.1
-
8
1249
AF025533.1
-
8
1393
AF031553.1
-
8
1329
AF031554.1
-
8
1329
AF031555.1
-
8
1329
AF031556.1
-
8
1329
AK223265.1
-
8
1380
AK290917.1
-
8
1408
BC028153.1
-
9
1412
BC104993.1
-
8
1346
8143996
BC112198.1
-
8
1346
8327666
EU814627.1
-
8
1327
EU814628.1
-
8
1327
KC918852.1
-
8
1371
KC918854.1
-
8
1371
KC918855.1
-
8
1371
KC918857.1
-
8
1371
KC918858.1
-
8
1371
KC918860.1
-
8
1371
KC918863.1
-
8
1371
U91928.1
-
8
1376
Gene Symbol: LILRA6
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
GenBank mRNA
KC918856.1
-
8
1371
KC918859.1
-
9
1371
Back To Top
More Information
Panther Classification:
Molecular Function -
immunoglobulin receptor superfamily
Gene Ontology Categories:
Function(s)
Process(es)
adaptive immune response
defense response
cell surface receptor signaling pathway
negative regulation of osteoclast differentiation
receptor activity
transmembrane signaling receptor activity
protein binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-5c7f9f6c68-zgjsk:80/10.239.161.116:80.
git-commit: b16971a8df2afcd8df684aafbc9bf1e9f45e42d4
git-url: git@github.com:thermofisher/magellan-core.git
git-branch: origin/release/2.6.0-2024.04.15-1.0