Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: UNC50 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_014044.5 | NP_054763.2 | 2 | 267 | ||
XM_005263913.4 | XP_005263970.1 | 2 | 363 | |||
XM_006712403.3 | XP_006712466.1 | 2 | 363 | |||
XM_006712404.2 | XP_006712467.1 | 2 | 277 | |||
GenBank mRNA | AF077038.1 | - | 2 | 219 | ||
AK222599.1 | - | 2 | 233 | |||
AL080115.1 | - | 2 | 271 | |||
AY017215.1 | - | 2 | 235 | |||
AY251534.1 | - | 1 | 128 | |||
BC017019.1 | - | 2 | 400 | 3888349 | ||
BC025992.2 | - | 1 | 1295 | 4830706 | ||
BC039078.1 | - | 1 | 1271 | 5166015 | ||
BC091481.1 | - | 2 | 257 | 30337350 | ||
CR533464.1 | - | 1 | 115 | |||
KU178586.1 | - | 1 | 115 | |||
KU178587.1 | - | 1 | 115 |