Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KIAA0101 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001029989.2 | NP_001025160.1 | 2 | 215 | ||
NM_014736.5 | NP_055551.1 | 2 | 215 | |||
NR_109934.1 | 2 | 271 | ||||
GenBank mRNA | AF529370.1 | - | 2 | 76 | ||
AK290748.1 | - | 2 | 146 | |||
AV757440.1 | - | 2 | 245 | |||
AY598324.1 | - | 2 | 76 | |||
BC005832.2 | - | 2 | 135 | 2988995 | ||
BC007101.1 | - | 2 | 142 | 4281110 | ||
BC016782.1 | - | 2 | 150 | 4109322 | ||
BF691807.1 | - | 2 | 216 | |||
BG502841.1 | - | 208 | ||||
D14657.1 | - | 2 | 137 | |||
DT220867.1 | - | 2 | 268 |