Search
Search

HSPB11| Gene Symbol: HSPB11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001316935.1 | NP_001303864.1 | 3, 4 | 292 | ||
| NM_016126.3 | NP_057210.2 | 3, 4 | 489 | |||
| GenBank mRNA | AF100747.1 | - | 3, 4 | 262 | ||
| AF416725.1 | - | 2, 3 | 216 | |||
| AF417114.1 | - | 2, 3 | 216 | |||
| AK225916.1 | - | 3, 4 | 240 | |||
| BC005245.1 | - | 3, 4 | 281 | 3838843 | ||
| HY179788.1 | - | 3, 4 | 293 | |||
| L32869.1 | - | 1, 2 | 64 | |||
| X79302.1 | - | 1, 2 | 64 | |||