Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HIRA | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_003325.3 | NP_003316.3 | 3 | 345 | ||
GenBank mRNA | AK289809.1 | - | 3 | 125 | ||
AK299950.1 | - | 4 | 284 | |||
AK302835.1 | - | 4 | 389 | |||
AK308642.1 | - | 4 | 284 | |||
BC032721.1 | - | 3 | 383 | |||
BC039835.1 | - | 3 | 340 | 5491361 | ||
CR456503.1 | - | 3 | 131 | |||
X75296.1 | - | 3 | 352 | |||
X77633.1 | - | 2 | 101 | |||
X81844.1 | - | 3 | 186 | |||
X89887.1 | - | 3 | 345 |