Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Custom Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Who We Serve
Biotech
Biopharma
CDMO
Lab Diagnostics
Industrial and Applied Sciences
Promotions
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Order Status
Quick Order
Promos
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other TPM3 SIRNA Assays ›
Gene Symbol:
TPM3
Gene Name:
tropomyosin 3
Gene Aliases:
CAPM1, CFTD, HEL-189, HEL-S-82p, NEM1, OK/SW-cl.5, TM-5, TM3, TM30, TM30nm, TM5, TPMsk3, TRK, hscp30
Chromosome Location:
Chr.1: 154155304 - 154192135 on Build GRCh38
UniGene ID:
Hs.535581
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
46535
Purification
Standard
HPLC
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: TPM3
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001043351.1
NP_001036816.1
2
366
NM_001043352.1
NP_001036817.1
2
366
NM_001043353.1
NP_001036818.1
2
366
NM_001278189.1
NP_001265118.1
2
366
NM_001278190.1
NP_001265119.1
2
366
NM_001278191.1
NP_001265120.1
2
212
NM_152263.3
NP_689476.2
3
464
NM_153649.3
NP_705935.1
2
366
NR_103461.1
2
366
GenBank mRNA
AF474157.1
-
2
236
AK026559.1
-
2
306
AK056921.1
-
2
223
AK092690.1
-
3
304
AK315504.1
-
2
325
AL537561.3
-
2
276
AY004867.1
-
2
247
BC000771.2
-
2
278
3506357
BC008407.1
-
3
430
4245271
BC008425.1
-
3
433
4246114
BC015403.1
-
2
281
4389911
BC072428.1
-
2
293
6472532
BF967161.1
-
2
179
BG706843.1
-
2
309
BI546300.1
-
2
277
BQ933569.1
-
2
333
BU956481.1
-
2
292
BX419658.2
-
2
279
BX648485.1
-
2
326
CR997359.1
-
2
257
DA209890.1
-
2
215
DA472065.1
-
2
366
DA900740.1
-
3
465
EU668324.1
-
3
304
FJ224335.1
-
2
326
KU178368.1
-
2
236
KU178369.1
-
2
236
KU178370.1
-
2
236
X04201.1
-
3
405
X04588.1
-
2
288
Back To Top
More Information
Panther Classification:
Molecular Function -
actin binding motor protein
Gene Ontology Categories:
Function(s)
Process(es)
movement of cell or subcellular component
muscle contraction
muscle filament sliding
molecular_function
actin binding
protein binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-green-659f68c6f4-wf7gw:80/100.66.73.223:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0