Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other B2M SIRNA Assays ›
          Gene Symbol:
          B2m
          Gene Name:
          beta-2 microglobulin
          Gene Aliases:
          -
          Chromosome Location:
          Chr.3: 114087287 - 114093311 on Build Rnor_6.0
          UniGene ID:
          Rn.1868
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          siRNA ID 46581

          Purification
          Size
          Availability Inventoried
          Catalog # AM16708
          Price 286.00
          Your Price
          Online offer: 277.65
          277.65
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: B2m
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_012512.2 NP_036644.1 2 113
          GenBank mRNA FM032790.1 - 2 113
          FQ210051.1 - 2 126
          FQ210252.1 - 2 127
          FQ210804.1 - 2 125
          FQ210906.1 - 2 126
          FQ211296.1 - 2 126
          FQ211419.1 - 2 125
          FQ212410.1 - 2 127
          FQ217926.1 - 2 124
          FQ218141.1 - 2 126
          FQ218251.1 - 2 126
          FQ218298.1 - 2 125
          FQ219157.1 - 2 128
          FQ219455.1 - 675
          FQ220256.1 - 2 122
          FQ221268.1 - 2 126
          FQ221287.1 - 2 122
          FQ221985.1 - 2 125
          FQ222352.1 - 2 123
          FQ225010.1 - 2 126
          FQ225051.1 - 2 130
          FQ225087.1 - 2 125
          FQ225088.1 - 2 127
          FQ225121.1 - 2 125
          FQ225141.1 - 2 125
          FQ225219.1 - 2 124
          FQ225292.1 - 2 126
          FQ225550.1 - 2 125
          FQ225647.1 - 2 125
          FQ225690.1 - 2 267
          FQ225912.1 - 2 125
          FQ226004.1 - 2 126
          FQ226258.1 - 2 128
          FQ226642.1 - 2 127
          FQ226723.1 - 2 125
          FQ227149.1 - 2 125
          FQ227291.1 - 2 131
          FQ227327.1 - 2 126
          FQ227454.1 - 2 126
          FQ227468.1 - 2 124
          FQ227656.1 - 2 126
          FQ227700.1 - 2 126
          FQ227950.1 - 2 124
          FQ227964.1 - 2 125
          FQ227995.1 - 2 121
          FQ228065.1 - 2 127
          FQ228188.1 - 2 126
          FQ228427.1 - 2 127
          FQ229482.1 - 2 118
          FQ229718.1 - 2 125
          FQ230202.1 - 2 126
          FQ230402.1 - 2 126
          FQ230977.1 - 2 125
          FQ231135.1 - 2 126
          FQ231182.1 - 2 126
          FQ231367.1 - 2 125
          FQ231378.1 - 2 125
          FQ231524.1 - 2 125
          FQ231602.1 - 2 126
          FQ231718.1 - 2 125
          FQ231856.1 - 2 125
          FQ231894.1 - 2 125
          FQ232233.1 - 2 126
          FQ232294.1 - 2 126
          FQ232460.1 - 2 125
          FQ232758.1 - 2 125
          FQ232818.1 - 2 125
          FQ232819.1 - 2 125
          FQ232937.1 - 2 125
          FQ232943.1 - 2 127
          FQ232957.1 - 2 123
          FQ233240.1 - 2 126
          FQ233290.1 - 2 125
          FQ233447.1 - 2 126
          FQ233505.1 - 2 125
          FQ233544.1 - 2 125
          FQ233567.1 - 2 130
          FQ233757.1 - 2 125
          FQ233797.1 - 2 124
          FQ233849.1 - 2 125
          FQ234348.1 - 2 126
          FQ234349.1 - 2 125
          FQ234524.1 - 2 123
          FQ234581.1 - 2 123
          FQ234840.1 - 2 125
          FQ234885.1 - 2 125
          FQ235105.1 - 2 125
          FQ235149.1 - 2 123
          FQ235229.1 - 2 125
          Y00441.1 - 2 100
          Y08531.1 - 2 105

          Back To Top

          More Information


          Panther Classification:

          Molecular Function -

          major histocompatibility complex protein

          Gene Ontology Categories:

          Function(s) Process(es)

          retina homeostasis
          positive regulation of T cell mediated cytotoxicity
          response to molecule of bacterial origin
          antigen processing and presentation of peptide antigen via MHC class I
          antigen processing and presentation of exogenous protein antigen via MHC class Ib, TAP-dependent
          T cell differentiation in thymus
          protein refolding
          response to drug
          response to cadmium ion
          iron ion homeostasis
          molecular_function
          identical protein binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-green-659f68c6f4-sg7j2:80/100.66.75.98:80.
          git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.43.0-2026.01.05-1.0