Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other B2M SIRNA Assays ›
          Gene Symbol:
          B2m
          Gene Name:
          beta-2 microglobulin
          Gene Aliases:
          -
          Chromosome Location:
          Chr.3: 114087287 - 114093311 on Build Rnor_6.0
          UniGene ID:
          Rn.1868
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          siRNA ID 46677

          Purification
          Size
          Availability Inventoried
          Catalog # AM16708
          Price 4,126.00
          Your Price
          Online offer:
          4126.0
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: B2m
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_012512.2 NP_036644.1 2 211
          GenBank mRNA FM032790.1 - 2 211
          FQ210051.1 - 2 224
          FQ210252.1 - 2 225
          FQ210804.1 - 2 223
          FQ210906.1 - 2 224
          FQ211296.1 - 2 224
          FQ211419.1 - 2 223
          FQ212410.1 - 2 225
          FQ217926.1 - 2 222
          FQ218141.1 - 2 224
          FQ218251.1 - 2 224
          FQ218298.1 - 2 223
          FQ219157.1 - 2 226
          FQ219455.1 - 773
          FQ221268.1 - 2 224
          FQ221287.1 - 2 220
          FQ221985.1 - 2 223
          FQ222352.1 - 2 221
          FQ225010.1 - 2 224
          FQ225051.1 - 2 228
          FQ225087.1 - 2 223
          FQ225088.1 - 2 225
          FQ225121.1 - 2 223
          FQ225141.1 - 2 223
          FQ225219.1 - 2 222
          FQ225292.1 - 2 224
          FQ225550.1 - 2 223
          FQ225647.1 - 2 223
          FQ225686.1 - 2 226
          FQ225690.1 - 2 365
          FQ225847.1 - 2 223
          FQ225912.1 - 2 223
          FQ226004.1 - 2 224
          FQ226258.1 - 2 226
          FQ226642.1 - 2 225
          FQ226723.1 - 2 223
          FQ227149.1 - 2 223
          FQ227291.1 - 2 229
          FQ227454.1 - 2 224
          FQ227468.1 - 2 222
          FQ227656.1 - 2 224
          FQ227700.1 - 2 224
          FQ227950.1 - 2 222
          FQ227964.1 - 2 223
          FQ227995.1 - 2 219
          FQ228065.1 - 2 225
          FQ228188.1 - 2 224
          FQ229482.1 - 2 216
          FQ229718.1 - 2 223
          FQ230202.1 - 2 224
          FQ230402.1 - 2 224
          FQ230977.1 - 2 223
          FQ231135.1 - 2 224
          FQ231182.1 - 2 224
          FQ231367.1 - 2 223
          FQ231378.1 - 2 223
          FQ231524.1 - 2 223
          FQ231602.1 - 2 224
          FQ231718.1 - 2 223
          FQ231856.1 - 2 223
          FQ231894.1 - 2 223
          FQ232233.1 - 2 224
          FQ232294.1 - 2 224
          FQ232460.1 - 2 223
          FQ232758.1 - 2 223
          FQ232818.1 - 2 223
          FQ232819.1 - 2 223
          FQ232937.1 - 2 223
          FQ232943.1 - 2 225
          FQ232957.1 - 2 221
          FQ233220.1 - 2 224
          FQ233240.1 - 2 224
          FQ233290.1 - 2 223
          FQ233447.1 - 2 224
          FQ233505.1 - 2 223
          FQ233544.1 - 2 223
          FQ233567.1 - 2 228
          FQ233757.1 - 2 223
          FQ233797.1 - 2 222
          FQ233849.1 - 2 223
          FQ234348.1 - 2 224
          FQ234349.1 - 2 223
          FQ234524.1 - 2 221
          FQ234581.1 - 2 221
          FQ234840.1 - 2 223
          FQ234885.1 - 2 223
          FQ235105.1 - 2 223
          FQ235149.1 - 2 221
          FQ235229.1 - 2 223
          Y00441.1 - 2 198
          Y08531.1 - 2 203

          Back To Top

          More Information


          Panther Classification:

          Molecular Function -

          major histocompatibility complex protein

          Gene Ontology Categories:

          Function(s) Process(es)

          retina homeostasis
          positive regulation of T cell mediated cytotoxicity
          response to molecule of bacterial origin
          antigen processing and presentation of peptide antigen via MHC class I
          antigen processing and presentation of exogenous protein antigen via MHC class Ib, TAP-dependent
          T cell differentiation in thymus
          protein refolding
          response to drug
          response to cadmium ion
          iron ion homeostasis
          molecular_function
          identical protein binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-green-659f68c6f4-st8h5:80/100.66.79.144:80.
          git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.43.0-2026.01.05-1.0