Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Cell Analysis
Antibodies
Mass Spectrometry
Cell Culture
Laboratory Instruments
Clinical and Diagnostics
Chromatography
Laboratory Equipment
Laboratory Supplies
Molecular Biology and Nucleic Acid Analysis
Sequence-Specific Nucleic Acid Products
See all product categories
Applications
Cell Culture and Transfection
Flow Cytometry
Cancer Research
Chromatography
Sequencing
PCR
Lab Solutions
Allergy Diagnostics
See all applications and techniques
Services
Cell Biology Services
Custom Services
Training Services
Enterprise Services
Lab Informatics Services
Financial and Leasing Services
Partnering and Licensing Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Food Safety Inspection Services
See all services
Help and Support
How to Order
Promotions and Online Offers
Contact Us
Change Location
Create a New Account
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Special Offers
Contact Us
Quick Order
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Contact Us
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Custom Products & Projects
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other ASCL1 SIRNA Assays ›
Gene Symbol:
Ascl1
Gene Name:
achaete-scute family bHLH transcription factor 1
Gene Aliases:
Mash1
Chromosome Location:
Chr.7: 28038662 - 28040504 on Build Rnor_6.0
UniGene ID:
Rn.32936
Species:
Rat
Species Specific ID (Flybase ID):
-
siRNA ID
46747
Purification
Standard
HPLC
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
4,126.00
Your Price
Online offer:
4126.0
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: Ascl1
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_022384.1
NP_071779.1
1
1195
GenBank mRNA
X53725.1
-
1
1195
Back To Top
More Information
Other Species Targeted
This siRNA also targets the following transcripts in Mouse: NM_008553, BC055748, AK141866, AK143210, M95603.
Panther Classification:
Molecular Function -
basic helix-loop-helix transcription factor
Gene Ontology Categories:
Function(s)
Process(es)
negative regulation of transcription from RNA polymerase II promoter
neuron migration
noradrenergic neuron development
noradrenergic neuron fate commitment
transcription, DNA-templated
regulation of transcription from RNA polymerase II promoter
Notch signaling pathway
regulation of mitotic cell cycle
pattern specification process
nervous system development
neuroblast fate determination
neuroblast proliferation
regulation of Notch signaling pathway
glial cell differentiation
response to lithium ion
regulation of gene expression
oligodendrocyte development
spinal cord association neuron differentiation
spinal cord oligodendrocyte cell differentiation
spinal cord oligodendrocyte cell fate specification
vestibular nucleus development
oligodendrocyte cell fate commitment
forebrain neuron differentiation
cerebral cortex GABAergic interneuron differentiation
commitment of neuronal cell to specific neuron type in forebrain
central nervous system neuron development
cerebral cortex development
neurogenesis
cell differentiation
neuron differentiation
regulation of epithelial cell differentiation
response to retinoic acid
regulation of cell proliferation
negative regulation of apoptotic process
positive regulation of neuron apoptotic process
negative regulation of neuron differentiation
positive regulation of neuron differentiation
positive regulation of Notch signaling pathway
positive regulation of cell cycle
negative regulation of transcription, DNA-templated
positive regulation of transcription from RNA polymerase II promoter
cell maturation
neuron fate commitment
neuron fate specification
neuron development
generation of neurons
oligodendrocyte differentiation
regulation of neurogenesis
positive regulation of neurogenesis
musculoskeletal movement, spinal reflex action
response to folic acid
subpallium neuron fate commitment
regulation of timing of subpallium neuron differentiation
olfactory pit development
ventral spinal cord interneuron fate commitment
lung neuroendocrine cell differentiation
stomach neuroendocrine cell differentiation
carotid body glomus cell differentiation
adrenal chromaffin cell differentiation
sympathetic ganglion development
response to epidermal growth factor
cellular response to magnetism
positive regulation of neural precursor cell proliferation
RNA polymerase II core promoter proximal region sequence-specific DNA binding
transcription factor activity, transcription factor binding
transcriptional repressor activity, RNA polymerase II core promoter proximal region sequence-specific binding
DNA binding
chromatin binding
double-stranded DNA binding
transcription factor activity, sequence-specific DNA binding
protein homodimerization activity
bHLH transcription factor binding
sequence-specific DNA binding
E-box binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-blue-55ff9ddd88-8jdkq:80/100.66.75.107:80.
git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.42.0-2026.01.03-1.0