Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RPS6KB2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_003952.2 | NP_003943.2 | 4 | 348 | ||
XM_005274164.1 | XP_005274221.1 | 3 | 296 | |||
XM_005274165.4 | XP_005274222.1 | 4 | 321 | |||
XM_006718655.3 | XP_006718718.1 | 4 | 329 | |||
XM_006718656.3 | XP_006718719.1 | 3 | 280 | |||
XM_006718657.1 | XP_006718720.1 | 3 | 262 | |||
XM_017018108.1 | XP_016873597.1 | 4 | 343 | |||
GenBank mRNA | AB016869.1 | - | 4 | 320 | ||
AB208827.1 | - | 2 | 762 | |||
AF076931.1 | - | 4 | 301 | |||
AF099739.1 | - | 4 | 305 | |||
AK297527.1 | - | 4 | 314 | |||
BC000094.2 | - | 4 | 321 | 3508140 | ||
BC006106.1 | - | 4 | 304 | 2959036 | ||
BC136564.1 | - | 4 | 322 | 9020554 |