Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture and Transfection
    • Chemicals
    • Chromatography
    • Electron Microscopes
    • Lab Plasticware and Supplies
    • Lab Centrifuges
    • Lab Solutions
    • Mass Spectrometers
    • Next Generation Sequencers
    • See all product categories
  • Applications
    • Brands
    • Industrial and Applied Sciences
    • Food and Beverage
    • Forensics
    • Lab Solutions
    • Life Sciences
    • Pharma and Biopharma
    • Biotechnology
    • Clinical and Diagnostics
    • Digital Solutions
    • See all applications
  • Services
    • Lab Instrument and Equipment Services
    • Enterprise Services
    • Custom Services
    • Training Services
    • Financial and Leasing Services
    • Enterprise Level Lab Informatics
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Cell Biology Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • Contact Us
    • Product Documentation
    • Knowledge Base and Product FAQs
    • Learning Centers
    • Supply Center
    • eProcurement Solutions
    • Lab Instrument and Equipment Support
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Special Offers
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other MT1A SIRNA Assays ›
          Gene Symbol:
          Mt1a Ttr
          Gene Name:
          metallothionein 1a
          transthyretin
          Gene Aliases:
          Mt, Mt1
          Lr1, Tbpa
          Chromosome Location:
          Chr.19: 11301991 - 11303007 on Build Rnor_6.0
          Chromosome Location:
          Chr.18: 15532963 - 15542180 on Build Rnor_6.0
          UniGene ID:
          Rn.54397
          Rn.1404
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          -
          siRNA ID 47027

          Purification
          Size
          Availability Inventoried
          Catalog # AM16708
          Price 4,126.00
          Your Price
          Online offer:
          4126.0
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: Mt1a
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_138826.4 NP_620181.1 3 290
          GenBank mRNA AF411318.1 - 1 124
          BC058442.1 - 3 295 6891030
          DQ255899.1 - 1 92
          FQ209933.1 - 3 291
          FQ210006.1 - 3 291
          FQ210086.1 - 3 291
          FQ210225.1 - 3 291
          FQ210276.1 - 3 291
          FQ210404.1 - 3 291
          FQ210574.1 - 3 291
          FQ210578.1 - 3 291
          FQ210592.1 - 3 291
          FQ210741.1 - 3 291
          FQ210753.1 - 3 291
          FQ210805.1 - 3 291
          FQ210924.1 - 3 291
          FQ210951.1 - 3 291
          FQ211146.1 - 3 291
          FQ211312.1 - 3 291
          FQ211350.1 - 3 291
          FQ217792.1 - 3 291
          FQ218077.1 - 3 291
          FQ218157.1 - 3 291
          FQ218299.1 - 3 291
          FQ218315.1 - 3 291
          FQ220411.1 - 3 291
          FQ221163.1 - 3 291
          FQ221173.1 - 3 291
          FQ221343.1 - 3 291
          FQ221498.1 - 3 291
          FQ221507.1 - 3 291
          FQ221511.1 - 3 291
          FQ221571.1 - 3 291
          FQ221639.1 - 3 291
          FQ221768.1 - 3 291
          FQ221874.1 - 3 291
          FQ222065.1 - 3 291
          FQ222070.1 - 3 291
          FQ222149.1 - 3 291
          FQ222220.1 - 3 291
          FQ222329.1 - 3 291
          FQ222341.1 - 3 291
          FQ222400.1 - 3 291
          FQ222420.1 - 3 291
          FQ222436.1 - 3 291
          FQ222448.1 - 3 291
          FQ222605.1 - 3 291
          FQ222614.1 - 3 290
          FQ222637.1 - 3 291
          FQ222674.1 - 3 291
          FQ222691.1 - 3 291
          FQ222786.1 - 3 291
          FQ222845.1 - 3 291
          FQ222891.1 - 3 291
          FQ222910.1 - 3 291
          FQ222929.1 - 3 291
          FQ222950.1 - 3 291
          FQ222957.1 - 3 290
          FQ223018.1 - 3 291
          FQ223045.1 - 3 291
          FQ223097.1 - 3 291
          FQ223170.1 - 3 291
          FQ223179.1 - 3 291
          FQ223313.1 - 3 291
          FQ223425.1 - 3 291
          FQ223452.1 - 3 291
          FQ228316.1 - 3 291
          FQ228334.1 - 3 291
          FQ228356.1 - 3 291
          FQ228476.1 - 3 291
          FQ228573.1 - 3 291
          FQ228587.1 - 3 291
          FQ228764.1 - 3 291
          FQ228980.1 - 3 291
          FQ229982.1 - 3 291
          J00750.1 - 3 290
          M24327.1 - 1 94
          Gene Symbol: Ttr
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          GenBank mRNA FQ210386.1 - 291

          Back To Top

          More Information


          Gene Ontology Categories:

          Function(s) Process(es)

          cellular metal ion homeostasis
          cellular zinc ion homeostasis
          nitric oxide mediated signal transduction
          response to zinc ion
          detoxification of copper ion
          negative regulation of neuron apoptotic process
          negative regulation of growth
          cellular response to chromate
          cellular response to cadmium ion
          cellular response to zinc ion
          copper ion binding
          zinc ion binding
          metal ion binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Quick Order
          • eProcurement
          • Supply Center
          • Order Status
          • Chemicals
          • India Mobile App
          • Government eMarketplace
          Support Plus Icon Minus Icon
          • Order Support
          • Training
          • Contact Us
          • Report a Site Issue
          • Instrument Management
          Resources Plus Icon Minus Icon
          • Product Selection Guides
          • Mobile & Desktop Apps
          • Webinars
          • Blog 
          • Social Media
          • New Products
          • Promotions
          • Shared Lists
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          India flag icon
          India

          Your items have has been added!


          Host server : magellan-search-blue-55ff9ddd88-8jdkq:80/100.66.75.107:80.
          git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.42.0-2026.01.03-1.0