Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Instrument Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Enterprise Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • Our Instagram
      Our Instagram
    • Our Facebook
      Our Facebook
    • Blog Behind the Bench
      Blog Behind the Bench
    • Customer Experience Center (CEC)
      Customer Experience Center (CEC)
    • Ecommerce Exclusives
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other MT1A SIRNA Assays ›
          Gene Symbol:
          Mt1a Ttr
          Gene Name:
          metallothionein 1a
          transthyretin
          Gene Aliases:
          Mt, Mt1
          Lr1, Tbpa
          Chromosome Location:
          Chr.19: 11301991 - 11303007 on Build Rnor_6.0
          Chromosome Location:
          Chr.18: 15532963 - 15542180 on Build Rnor_6.0
          UniGene ID:
          Rn.54397
          Rn.1404
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          -
          siRNA ID 47113

          Purification
          Size
          Availability Inventoried
          Catalog # AM16708
          Price 286.00
          Your Price
          Online offer: 277.65
          277.65
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: Mt1a
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_138826.4 NP_620181.1 3 368
          GenBank mRNA AF411318.1 - 1 202
          BC058442.1 - 3 373 6891030
          DQ255899.1 - 1 170
          FQ209933.1 - 3 369
          FQ210006.1 - 3 369
          FQ210086.1 - 3 369
          FQ210225.1 - 3 369
          FQ210276.1 - 3 369
          FQ210404.1 - 3 369
          FQ210574.1 - 3 369
          FQ210592.1 - 3 369
          FQ210741.1 - 3 369
          FQ210753.1 - 3 369
          FQ210805.1 - 3 369
          FQ210924.1 - 3 369
          FQ210951.1 - 3 369
          FQ211312.1 - 3 369
          FQ211350.1 - 3 369
          FQ217792.1 - 3 369
          FQ218077.1 - 3 369
          FQ218157.1 - 3 369
          FQ218299.1 - 3 369
          FQ218315.1 - 3 369
          FQ218775.1 - 3 369
          FQ220411.1 - 3 369
          FQ221163.1 - 3 369
          FQ221173.1 - 3 369
          FQ221343.1 - 3 369
          FQ221498.1 - 3 369
          FQ221507.1 - 3 369
          FQ221511.1 - 3 369
          FQ221516.1 - 3 368
          FQ221571.1 - 3 369
          FQ221639.1 - 3 369
          FQ221768.1 - 3 369
          FQ221874.1 - 3 369
          FQ222065.1 - 3 369
          FQ222070.1 - 3 369
          FQ222149.1 - 3 369
          FQ222220.1 - 3 369
          FQ222329.1 - 3 369
          FQ222341.1 - 3 369
          FQ222400.1 - 3 369
          FQ222420.1 - 3 369
          FQ222436.1 - 3 369
          FQ222448.1 - 3 369
          FQ222605.1 - 3 369
          FQ222614.1 - 3 368
          FQ222637.1 - 3 369
          FQ222674.1 - 3 369
          FQ222691.1 - 3 369
          FQ222845.1 - 3 369
          FQ222891.1 - 3 369
          FQ222907.1 - 3 369
          FQ222910.1 - 3 369
          FQ222929.1 - 3 369
          FQ222950.1 - 3 369
          FQ222957.1 - 3 368
          FQ223018.1 - 3 369
          FQ223045.1 - 3 369
          FQ223097.1 - 3 369
          FQ223170.1 - 3 370
          FQ223179.1 - 3 369
          FQ223313.1 - 3 369
          FQ223425.1 - 3 369
          FQ223452.1 - 3 369
          FQ228316.1 - 3 369
          FQ228334.1 - 3 369
          FQ228356.1 - 3 369
          FQ228476.1 - 3 369
          FQ228561.1 - 3 369
          FQ228573.1 - 3 369
          FQ228587.1 - 3 369
          FQ228764.1 - 3 369
          FQ228920.1 - 3 369
          FQ228980.1 - 3 369
          FQ229826.1 - 3 369
          FQ229982.1 - 3 369
          J00750.1 - 3 368
          M24327.1 - 1 172
          Gene Symbol: Ttr
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          GenBank mRNA FQ210386.1 - 369

          Back To Top

          More Information


          Gene Ontology Categories:

          Function(s) Process(es)

          cellular metal ion homeostasis
          cellular zinc ion homeostasis
          nitric oxide mediated signal transduction
          response to zinc ion
          detoxification of copper ion
          negative regulation of neuron apoptotic process
          negative regulation of growth
          cellular response to chromate
          cellular response to cadmium ion
          cellular response to zinc ion
          copper ion binding
          zinc ion binding
          metal ion binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Find Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Brasil flag icon
          Brasil

          Your items have has been added!


          Host server : magellan-search-blue-55ff9ddd88-lffdw:80/100.66.79.31:80.
          git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.42.0-2026.01.03-1.0