Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Psip1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_175765.2 | NP_786941.1 | 1, 2 | 175 | ||
XM_006238353.2 | XP_006238415.1 | 2, 3 | 399 | |||
XM_006238354.2 | XP_006238416.1 | 2, 3 | 398 | |||
XM_008763779.1 | XP_008762001.1 | 2, 3 | 398 | |||
GenBank mRNA | AB285525.1 | - | 78 | |||
AF339084.1 | - | 1, 2 | 175 | |||
AF372090.1 | - | 1, 2 | 175 | |||
BC093606.1 | - | 2, 3 | 333 | 7317952 |