Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Who We Serve
Biotech
Biopharma
CDMO
Lab Diagnostics
Industrial and Applied Sciences
Promotions
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Order Status
Quick Order
Promos
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other IL6 SIRNA Assays ›
Gene Symbol:
Il6
Gene Name:
interleukin 6
Gene Aliases:
ILg6, Ifnb2
Chromosome Location:
Chr.4: 3043231 - 3047807 on Build Rnor_6.0
UniGene ID:
Rn.9873
Species:
Rat
Species Specific ID (Flybase ID):
-
siRNA ID
47845
Purification
Standard
HPLC
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
4,126.00
Your Price
Online offer:
4126.0
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: Il6
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_012589.2
NP_036721.1
2
229
GenBank mRNA
M26744.1
-
2
229
Back To Top
More Information
Gene Ontology Categories:
Function(s)
Process(es)
neutrophil apoptotic process
response to yeast
myeloid cell homeostasis
hepatic immune response
positive regulation of acute inflammatory response
negative regulation of protein kinase activity
acute-phase response
inflammatory response
aging
positive regulation of cell proliferation
negative regulation of cell proliferation
regulation of cell shape
response to heat
response to cold
response to wounding
response to mechanical stimulus
response to bacterium
regulation of vascular endothelial growth factor production
positive regulation of gene expression
response to auditory stimulus
response to organic cyclic compound
cell growth
cytokine-mediated signaling pathway
response to caffeine
endocrine pancreas development
neuron projection development
response to nutrient levels
response to peptidoglycan
response to lipopolysaccharide
positive regulation of chemokine production
positive regulation of interleukin-6 production
response to insulin
negative regulation of collagen biosynthetic process
positive regulation of peptidyl-serine phosphorylation
positive regulation of protein import into nucleus, translocation
response to cytokine
wound healing
positive regulation of T cell proliferation
T cell activation
regulation of cell proliferation
response to drug
positive regulation of tyrosine phosphorylation of Stat3 protein
glucose homeostasis
defense response to protozoan
regulation of apoptotic process
positive regulation of apoptotic process
negative regulation of apoptotic process
negative regulation of cysteine-type endopeptidase activity involved in apoptotic process
response to amino acid
positive regulation of MAPK cascade
negative regulation of chemokine biosynthetic process
regulation of circadian sleep/wake cycle, non-REM sleep
positive regulation of nitric oxide biosynthetic process
cell redox homeostasis
positive regulation of T-helper 2 cell differentiation
positive regulation of neuron differentiation
negative regulation of gluconeogenesis
positive regulation of translation
positive regulation of DNA replication
positive regulation of transcription, DNA-templated
positive regulation of transcription from RNA polymerase II promoter
positive regulation of JAK-STAT cascade
response to antibiotic
muscle cell cellular homeostasis
bone remodeling
negative regulation of hormone secretion
negative regulation of muscle organ development
positive regulation of smooth muscle cell proliferation
positive regulation of epithelial cell proliferation
negative regulation of cytokine secretion
positive regulation of peptidyl-tyrosine phosphorylation
defense response to Gram-negative bacterium
defense response to Gram-positive bacterium
positive regulation of B cell activation
positive regulation of immunoglobulin secretion
positive regulation of sequence-specific DNA binding transcription factor activity
positive regulation of NF-kappaB transcription factor activity
response to glucocorticoid
response to calcium ion
response to electrical stimulus
defense response to virus
positive regulation of protein kinase B signaling
positive regulation of transmission of nerve impulse
branching involved in salivary gland morphogenesis
epithelial cell proliferation involved in salivary gland morphogenesis
glucagon secretion
interleukin-6-mediated signaling pathway
cellular response to hydrogen peroxide
positive regulation of ERK1 and ERK2 cascade
cellular response to lipopolysaccharide
cellular response to interleukin-1
cellular response to tumor necrosis factor
cellular response to dexamethasone stimulus
T-helper 17 cell lineage commitment
negative regulation of neuron death
receptor binding
cytokine activity
interleukin-6 receptor binding
growth factor activity
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-green-55658555d6-72rkq:80/100.66.79.224:80.
git-commit: ec8e7df5f8fec8d765bd419205f1b5046a016d37
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.41.0-Offline