Search
Search

Atp1a1| Gene Symbol: Atp1a1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_012504.1 | NP_036636.1 | 16, 17 | 2538 | ||
| GenBank mRNA | BC061968.1 | - | 16, 17 | 2549 | 5598014 | |
| FQ213537.1 | - | 1, 2 | 141 | |||
| M14511.1 | - | 16, 17 | 2538 | |||
| M28647.1 | - | 16, 17 | 2561 | |||
| X05882.1 | - | 16, 17 | 2315 | |||