Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TNFSF10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001190942.1 | NP_001177871.1 | 2 | 269 | ||
NM_003810.3 | NP_003801.1 | 2 | 269 | |||
NR_033994.1 | 2 | 269 | ||||
GenBank mRNA | AK296085.1 | - | 2 | 237 | ||
AK312742.1 | - | 2 | 237 | |||
AY312579.1 | - | 2 | 33 | |||
BC009795.1 | - | 2 | 166 | |||
BC032722.1 | - | 2 | 221 | 5504670 | ||
BT019564.1 | - | 2 | 146 | |||
CR456895.1 | - | 2 | 146 | |||
DA041872.1 | - | 2 | 229 | |||
DA969653.1 | - | 2 | 269 | |||
DQ848564.1 | - | 2 | 146 | |||
EU183231.1 | - | 2 | 146 | |||
U37518.1 | - | 2 | 233 | |||
U57059.1 | - | 2 | 236 |