Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FUBP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001303433.1 | NP_001290362.1 | 2 | 357 | ||
NM_003902.4 | NP_003893.2 | 2 | 357 | |||
NR_130152.1 | 2 | 357 | ||||
XM_011542391.2 | XP_011540693.1 | 2 | 228 | |||
XM_011542392.2 | XP_011540694.1 | 2 | 262 | |||
XM_011542393.2 | XP_011540695.1 | 2 | 262 | |||
XM_017002739.1 | XP_016858228.1 | 3 | 250 | |||
XM_017002740.1 | XP_016858229.1 | 2 | 228 | |||
XM_017002741.1 | XP_016858230.1 | 3 | 309 | |||
XM_017002742.1 | XP_016858231.1 | 2 | 262 | |||
XM_017002743.1 | XP_016858232.1 | 2 | 266 | |||
XM_017002744.1 | XP_016858233.1 | 2 | 266 | |||
GenBank mRNA | AK300032.1 | - | 2 | 225 | ||
AK301581.1 | - | 2 | 225 | |||
AK303568.1 | - | 2 | 224 | |||
BC010083.2 | - | 2 | 192 | |||
BC017247.2 | - | 2 | 221 | 4891583 | ||
BQ649331.1 | - | 2 | 245 | |||
DA113820.1 | - | 2 | 357 | |||
U05040.2 | - | 2 | 182 |