Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Htr4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012853.1 | NP_036985.1 | 4 | 506 | ||
XM_006254793.2 | XP_006254855.1 | 5 | 1045 | |||
XM_008772089.1 | XP_008770311.1 | 3 | 2171 | |||
XM_008772090.1 | XP_008770312.1 | 3 | 2171 | |||
GenBank mRNA | AJ011370.1 | - | 5 | 586 | ||
AM712909.1 | - | 4 | 486 | |||
AM712910.1 | - | 4 | 486 | |||
AM712911.1 | - | 4 | 486 | |||
U20906.1 | - | 4 | 506 | |||
U20907.1 | - | 4 | 506 | |||
Z48153.1 | - | 2 | 204 |
Set Membership: |
G-protein coupled receptor |