Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Custom Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Who We Serve
Biotech
Biopharma
CDMO
Lab Diagnostics
Industrial and Applied Sciences
Promotions
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Order Status
Quick Order
Promos
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other TP63 SIRNA Assays ›
Gene Symbol:
TP63
Gene Name:
tumor protein p63
Gene Aliases:
AIS, B(p51A), B(p51B), EEC3, KET, LMS, NBP, OFC8, RHS, SHFM4, TP53CP, TP53L, TP73L, p40, p51, p53CP, p63, p73H, p73L
Chromosome Location:
Chr.3: 189597115 - 189897279 on Build GRCh38
UniGene ID:
Hs.137569
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
4893
Purification
Standard
HPLC
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: TP63
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001114978.1
NP_001108450.1
6, 7
979
NM_001114979.1
NP_001108451.1
6, 7
979
NM_001114980.1
NP_001108452.1
4, 5
749
NM_001114981.1
NP_001108453.1
4, 5
749
NM_001114982.1
NP_001108454.1
4, 5
749
NM_003722.4
NP_003713.3
6, 7
979
XM_005247843.3
XP_005247900.1
6, 7
1426
XM_005247844.4
XP_005247901.1
6, 7
1142
XM_011513251.2
XP_011511553.1
7, 8
1020
XM_011513252.2
XP_011511554.1
6, 7
952
XM_017007387.1
XP_016862876.1
4, 5
757
GenBank mRNA
AB010153.1
-
4, 5
666
AB016072.1
-
6, 7
917
AB016073.1
-
6, 7
917
AB042841.1
-
3, 4
353
AF075428.1
-
5, 6
773
AF075429.1
-
4, 5
608
AF075430.1
-
5, 6
773
AF075431.1
-
4, 5
608
AF075432.1
-
5, 6
773
AF075433.1
-
4, 5
608
AF091627.1
-
4, 5
733
AF116771.1
-
6, 7
917
AJ315499.1
-
3, 4
434
AK304127.1
-
4, 5
749
BC039815.1
-
6, 7
979
5552611
GQ202689.1
-
4, 5
747
GQ202690.1
-
4, 5
747
KP881229.1
-
4, 5
613
KP881230.1
-
4, 5
613
KP881231.1
-
3, 4
358
KU178435.1
-
6, 7
890
KU178436.1
-
6, 7
890
Y16961.1
-
6, 7
917
Back To Top
More Information
Panther Classification:
Molecular Function -
P53-like transcription factor
Gene Ontology Categories:
Function(s)
Process(es)
negative regulation of transcription from RNA polymerase II promoter
replicative cell aging
skeletal system development
establishment of planar polarity
positive regulation of mesenchymal cell proliferation
epithelial cell development
chromatin remodeling
transcription from RNA polymerase II promoter
apoptotic process
DNA damage response, signal transduction by p53 class mediator resulting in transcription of p21 class mediator
Notch signaling pathway
spermatogenesis
ectoderm and mesoderm interaction
proximal/distal pattern formation
response to X-ray
multicellular organism aging
response to gamma radiation
epidermal cell division
regulation of epidermal cell division
positive regulation of keratinocyte proliferation
keratinocyte differentiation
embryonic limb morphogenesis
polarized epithelial cell differentiation
hair follicle morphogenesis
mitotic G1 DNA damage checkpoint
cellular response to UV
post-anal tail morphogenesis
odontogenesis of dentin-containing tooth
intrinsic apoptotic signaling pathway in response to DNA damage by p53 class mediator
regulation of apoptotic process
negative regulation of apoptotic process
regulation of cysteine-type endopeptidase activity involved in apoptotic process
regulation of neuron apoptotic process
skin morphogenesis
keratinocyte proliferation
negative regulation of keratinocyte differentiation
positive regulation of osteoblast differentiation
positive regulation of Notch signaling pathway
negative regulation of transcription, DNA-templated
positive regulation of transcription, DNA-templated
positive regulation of transcription from RNA polymerase II promoter
sympathetic nervous system development
smooth muscle tissue development
female genitalia morphogenesis
protein homotetramerization
neuron apoptotic process
urinary bladder development
cloacal septation
prostatic bud formation
squamous basal epithelial stem cell differentiation involved in prostate gland acinus development
establishment of skin barrier
positive regulation of protein insertion into mitochondrial membrane involved in apoptotic signaling pathway
regulation of signal transduction by p53 class mediator
positive regulation of fibroblast apoptotic process
negative regulation of mesoderm development
transcription factor activity, transcription factor binding
transcriptional activator activity, RNA polymerase II core promoter proximal region sequence-specific binding
p53 binding
DNA binding
chromatin binding
damaged DNA binding
double-stranded DNA binding
transcription factor activity, sequence-specific DNA binding
protein binding
identical protein binding
sequence-specific DNA binding
transcription regulatory region DNA binding
metal ion binding
WW domain binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-green-659f68c6f4-wf7gw:80/100.66.73.223:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0