Hamburger Menu Button
Thermo Fisher Scientific Logo
登入
還沒有帳戶嗎 ?
立即註冊
產品
Antibodies
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Cell Culture Media
Chemicals
Western Blot Products
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Greener Products
查看所有產品類別
應用
生物製程
細胞培養和轉染
食品和飲料
實驗室解決方案
製藥和生物製藥
即時定量PCR
半導體分析
臨床和診斷
數位解決方案
分子光譜、同位素與元素分析
查看所有應用與技術
服務
360° CDMO 和 CRO 服務
CDMO 服务
临床试验 CRO 服务
客製化服務
企業服務
金融與租賃服務
儀器服務
實驗室資訊學
授權與商業供應
培訓服務
查看所有服務
支援
線上訂購協助
數位解決方案
產品支持
技術相關資訊
培訓與教育
查看所有協助與支援
熱門商品
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
特價商品
聯絡我們
快速訂購
訂單狀態與貨物追蹤
文件和證書
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
訂單查詢
快速訂購
登入
登入
還沒有帳戶嗎 ?
立即註冊
帳戶
訂單
Connect: Lab, Data, Apps
客製化產品與項目
儀器管理
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other ICAM1 SIRNA Assays ›
Gene Symbol:
Icam1
Gene Name:
intercellular adhesion molecule 1
Gene Aliases:
CD54, ICAM
Chromosome Location:
Chr.8: 22035287 - 22047049 on Build Rnor_6.0
UniGene ID:
Rn.12
Species:
Rat
Species Specific ID (Flybase ID):
-
siRNA ID
48950
Purification
Standard
HPLC
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
286.00
Your Price
Online offer:
277.65
277.65
Check your price ›
新增到購物車
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: Icam1
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_012967.1
NP_037099.1
7
1609
GenBank mRNA
BC081837.1
-
7
1621
7122397
D00913.1
-
7
1609
FQ228241.1
-
7
1629
Back To Top
More Information
Other Species Targeted
This siRNA also targets the following transcripts in Mouse: NM_010493, AK149945, AK152530, AK149781, CT010302, and others.
Gene Ontology Categories:
Function(s)
Process(es)
ovarian follicle development
response to hypoxia
response to amphetamine
T cell activation via T cell receptor contact with antigen bound to MHC molecule on antigen presenting cell
acute inflammatory response to antigenic stimulus
T cell antigen processing and presentation
positive regulation of cellular extravasation
cell adhesion
leukocyte cell-cell adhesion
cell aging
sensory perception of sound
regulation of cell shape
response to ionizing radiation
response to sulfur dioxide
response to organic cyclic compound
single organismal cell-cell adhesion
membrane to membrane docking
regulation of cell adhesion
positive regulation of actin filament polymerization
cellular response to nutrient levels
response to lipopolysaccharide
cell adhesion mediated by integrin
response to gonadotropin
response to drug
response to amino acid
positive regulation of GTPase activity
adhesion of symbiont to host
positive regulation of nitric oxide biosynthetic process
response to ethanol
positive regulation of vasoconstriction
response to copper ion
receptor-mediated virion attachment to host cell
positive regulation of peptidyl-tyrosine phosphorylation
leukocyte migration
positive regulation of NF-kappaB transcription factor activity
negative regulation of calcium ion transport
establishment of endothelial barrier
positive regulation of ERK1 and ERK2 cascade
cellular response to lipopolysaccharide
cellular response to organic substance
cellular response to alkaloid
cellular response to glucose stimulus
cellular response to interleukin-1
cellular response to tumor necrosis factor
cellular response to hypoxia
establishment of Sertoli cell barrier
regulation of ruffle assembly
negative regulation of extrinsic apoptotic signaling pathway via death domain receptors
negative regulation of endothelial cell apoptotic process
integrin binding
protein complex binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-blue-55ff9ddd88-8jdkq:80/100.66.75.107:80.
git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.42.0-2026.01.03-1.0