Search
Search

TNFSF10| Gene Symbol: TNFSF10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_003810.3 | NP_003801.1 | 3 | 422 | ||
| GenBank mRNA | AK312742.1 | - | 3 | 390 | ||
| BC009795.1 | - | 3 | 319 | |||
| BC032722.1 | - | 3 | 374 | 5504670 | ||
| BT019564.1 | - | 3 | 299 | |||
| CR456895.1 | - | 3 | 299 | |||
| DA969653.1 | - | 3 | 422 | |||
| U37518.1 | - | 3 | 386 | |||
| U57059.1 | - | 3 | 389 | |||