Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Si | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_013061.1 | NP_037193.1 | 3 | 281 | ||
GenBank mRNA | L25926.1 | - | 3 | 281 |