Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
CDMO and Clinical Trial Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Instrument Management
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other CREM SIRNA Assays ›
Gene Symbol:
Crem
Gene Name:
cAMP responsive element modulator
Gene Aliases:
Icer
Chromosome Location:
Chr.17: 57021704 - 57090888 on Build Rnor_6.0
UniGene ID:
Rn.10251
Species:
Rat
Species Specific ID (Flybase ID):
-
siRNA ID
49133
Purification
HPLC
Standard
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: Crem
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001110860.2
NP_001104330.1
9, 10
1281
NM_001271101.1
NP_001258030.1
9, 10
1281
NM_001271102.1
NP_001258031.1
2, 3
235
NM_001271245.1
NP_001258174.1
3, 4
317
NM_001271246.1
NP_001258175.1
3, 4
379
NM_001271247.1
NP_001258176.1
3, 4
425
NM_001271248.1
NP_001258177.1
2, 3
343
NM_017334.2
NP_059030.2
3, 4
271
XM_006254049.2
XP_006254111.1
8, 9
1239
XM_006254050.2
XP_006254112.1
8, 9
1239
XM_006254051.2
XP_006254113.1
8, 9
1065
XM_006254052.2
XP_006254114.1
7, 8
1397
XM_006254053.2
XP_006254115.1
8, 9
1019
XM_006254054.2
XP_006254116.1
7, 8
1119
XM_006254055.2
XP_006254117.1
7, 8
983
XM_006254056.2
XP_006254118.1
7, 8
983
XM_006254057.2
XP_006254119.1
8, 9
1086
XM_006254058.2
XP_006254120.1
7, 8
1050
XM_006254059.2
XP_006254121.1
7, 8
1050
XM_006254060.2
XP_006254122.1
6, 7
1250
XM_006254061.2
XP_006254123.1
6, 7
1250
XM_006254062.2
XP_006254124.1
5, 6
1214
XM_006254063.2
XP_006254125.1
6, 7
969
XM_006254064.2
XP_006254126.1
7, 8
830
XM_006254065.2
XP_006254127.1
7, 8
830
XM_006254066.2
XP_006254128.1
6, 7
794
XM_006254067.2
XP_006254129.1
6, 7
794
XM_006254068.2
XP_006254130.1
4, 5
1025
XM_006254069.2
XP_006254131.1
3, 4
658
XM_006254070.2
XP_006254132.1
2, 3
1143
XM_008771691.1
XP_008769913.1
3, 4
704
XM_008771692.1
XP_008769914.1
3, 4
300
XM_008771693.1
XP_008769915.1
2, 3
942
XM_008771694.1
XP_008769916.1
2, 3
279
XM_008771695.1
XP_008769917.1
2, 3
258
GenBank mRNA
AB031423.1
-
3, 4
300
BC078899.1
-
9, 10
1281
7133070
CO556486.1
-
2, 3
292
FM033648.1
-
3, 4
415
FM089481.1
-
3, 4
380
FQ213544.1
-
2, 3
235
FQ219343.1
-
2, 3
232
FQ220921.1
-
2, 3
234
S66024.1
-
3, 4
268
U04835.1
-
3, 4
309
Z15158.1
-
7, 8
874
Back To Top
More Information
Other Species Targeted
This siRNA also targets the following transcripts in Mouse: NM_001110857, NM_001271506, NM_001110858, NM_001271503, NM_001110854, and others.
Gene Ontology Categories:
Function(s)
Process(es)
glucose metabolic process
transcription, DNA-templated
regulation of transcription, DNA-templated
fatty acid metabolic process
glycosphingolipid metabolic process
spermatogenesis
cAMP-mediated signaling
circadian regulation of gene expression
regulation of circadian rhythm
negative regulation of transcription, DNA-templated
positive regulation of transcription from RNA polymerase II promoter
retinoic acid receptor signaling pathway
response to cAMP
core promoter sequence-specific DNA binding
DNA binding
transcription factor activity, sequence-specific DNA binding
sequence-specific DNA binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-75dc5564b9-zvggx:80/10.239.163.62:80.
git-commit: 1b386f5412c20333c1bb415eea69346c63d8df9e
git-url: git@github.com:thermofisher/magellan-core.git
git-branch: origin/release/2.7.0-2024.04.17-1.0