Search
Search

Anxa5| Gene Symbol: Anxa5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_013132.1 | NP_037264.1 | 4, 5 | 258 | ||
| XM_006232268.2 | XP_006232330.1 | 4, 5 | 393 | |||
| GenBank mRNA | AF051895.1 | - | 4, 5 | 313 | ||
| BC081855.1 | - | 4, 5 | 311 | 7192977 | ||
| FQ213209.1 | - | 5, 6 | 388 | |||
| FQ229127.1 | - | 4, 5 | 246 | |||
| M21730.1 | - | 4, 5 | 258 | |||