Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mc5r | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_013182.2 | NP_037314.1 | 1 | 969 | ||
GenBank mRNA | L27081.1 | - | 1 | 1181 |
Set Membership: |
G-protein coupled receptor |