siRNA Details
Gene Symbol: Tbxa2r | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_017054.1 | NP_058750.1 | 2 | 1103 | ||
XM_006240846.1 | XP_006240908.1 | 2 | 1261 | |||
XM_006240847.1 | XP_006240909.1 | 1 | 813 | |||
GenBank mRNA | D21158.1 | - | 2 | 1012 | ||
D32080.1 | - | 2 | 1103 |

More Information
Other Species Targeted

Set Membership: |
![]() |
Panther Classification:
