Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Western Blot Products
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Food and Beverage
    • Lab Solutions
    • Pharma and Biopharma
    • Real-Time PCR
    • Semiconductor Analysis
    • Clinical and Diagnostics
    • Digital Solutions
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • See all services
  • Help and Support
    • Order Help
    • Digital Solutions
    • Product Support
    • Technical Information
    • Training and Education
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Special Offers
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other ADORA1 SIRNA Assays ›
          Gene Symbol:
          Adora1
          Gene Name:
          adenosine A1 receptor
          Gene Aliases:
          -
          Chromosome Location:
          Chr.13: 51042246 - 51076856 on Build Rnor_6.0
          UniGene ID:
          Rn.32078
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          siRNA ID 49929

          Purification
          Size
          Availability Inventoried
          Catalog # AM16708
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: Adora1
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_017155.2 NP_058851.2 2 677
          XM_006249859.2 XP_006249921.1 3 968
          XM_006249860.2 XP_006249922.1 3 743
          XM_006249861.2 XP_006249923.1 3 907
          XM_008769552.1 XP_008767774.1 3 1130
          XM_008769553.1 XP_008767775.1 3 1069
          GenBank mRNA AB001089.1 - 2 614
          AF042079.1 - 2 645
          M69045.1 - 2 677
          Y12519.1 - 2 1029

          Back To Top

          More Information


          Other Species Targeted

          This siRNA also targets the following transcripts in Mouse: U05671.

          Set Membership:

          G-protein coupled receptor

          Panther Classification:

          Molecular Function -

          G-protein coupled receptor

          Gene Ontology Categories:

          Function(s) Process(es)

          activation of MAPKK activity
          temperature homeostasis
          response to hypoxia
          adenosine receptor signaling pathway
          regulation of respiratory gaseous exchange by neurological system process
          negative regulation of acute inflammatory response
          negative regulation of leukocyte migration
          positive regulation of peptide secretion
          regulation of glomerular filtration
          protein targeting to membrane
          G-protein coupled receptor signaling pathway
          adenylate cyclase-inhibiting G-protein coupled receptor signaling pathway
          negative regulation of cell proliferation
          negative regulation of glutamate secretion
          lipid catabolic process
          negative regulation of synaptic transmission, GABAergic
          positive regulation of nucleoside transport
          negative regulation of neurotrophin production
          positive regulation of protein dephosphorylation
          negative regulation of renal sodium excretion
          regulation of vasodilation
          negative regulation of circadian sleep/wake cycle, sleep
          negative regulation of circadian sleep/wake cycle, non-REM sleep
          negative regulation of apoptotic process
          positive regulation of potassium ion transport
          positive regulation of epidermal growth factor-activated receptor activity
          negative regulation of blood pressure
          positive regulation of blood pressure
          negative regulation of heart contraction
          negative regulation of vasodilation
          negative regulation of hormone secretion
          regulation of synaptic plasticity
          negative regulation of inflammatory response
          cognition
          detection of temperature stimulus involved in sensory perception of pain
          negative regulation of lipid catabolic process
          regulation of sensory perception of pain
          negative regulation of synaptic transmission, glutamatergic
          regulation of cardiac muscle contraction
          negative regulation of cardiac muscle contraction
          excitatory postsynaptic potential
          relaxation of vascular smooth muscle
          negative regulation of mucus secretion
          negative regulation of long term synaptic depression
          G-protein coupled adenosine receptor activity
          G-protein coupled receptor binding
          purine nucleoside binding
          phospholipase C activity
          protein binding
          G-protein beta/gamma-subunit complex binding
          heterotrimeric G-protein binding
          protein heterodimerization activity

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Taiwan flag icon
          Taiwan

          Your items have has been added!


          Host server : magellan-search-blue-55ff9ddd88-5btgp:80/100.66.77.235:80.
          git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.42.0-2026.01.03-1.0