Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Esp3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001251916.1 | NP_001238845.1 | 3 | 839 | ||
GenBank mRNA | AB306981.1 | - | 3 | 839 |