Search Thermo Fisher Scientific
Gene Symbol: Gm20172 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001204913.1 | NP_001191842.1 | 1 | 959 | ||
GenBank mRNA | AK084043.1 | - | 1 | 959 | ||
AK148788.1 | - | 1 | 959 |