Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Cell Analysis
Antibodies
Mass Spectrometry
Cell Culture
Laboratory Instruments
Clinical and Diagnostics
Chromatography
Laboratory Equipment
Laboratory Supplies
Molecular Biology and Nucleic Acid Analysis
Sequence-Specific Nucleic Acid Products
See all product categories
Applications
Cell Culture and Transfection
Flow Cytometry
Cancer Research
Chromatography
Sequencing
PCR
Lab Solutions
Allergy Diagnostics
See all applications and techniques
Services
Cell Biology Services
Custom Services
Training Services
Lab Informatics Services
Financial and Leasing Services
Partnering and Licensing Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Food Safety Inspection Services
See all services
Help and Support
How to Order
Promotions and Online Offers
Contact Us
Change Location
Create a New Account
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Who We Serve
Biotech
Biopharma
CDMO
Lab Diagnostics
Industrial and Applied Sciences
Special Offers
Contact Us
Quick Order
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Contact Us
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Custom Products & Projects
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other ERBB3 SIRNA Assays ›
Gene Symbol:
Erbb3
Gene Name:
erb-b2 receptor tyrosine kinase 3
Gene Aliases:
nuc-ErbB3
Chromosome Location:
Chr.7: 2991016 - 3010312 on Build Rnor_6.0
UniGene ID:
Rn.10228
Species:
Rat
Species Specific ID (Flybase ID):
-
siRNA ID
50076
Purification
Standard
HPLC
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
286.00
Your Price
Online offer:
277.65
277.65
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: Erbb3
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_017218.3
NP_058914.2
28
3722
GenBank mRNA
GU598254.1
-
6
1181
U29339.2
-
28
3722
Back To Top
More Information
Other Species Targeted
This siRNA also targets the following transcripts in Human: NM_001982, BC082992, AK295650, BC002706, M34309, and others.
This siRNA also targets the following transcripts in Mouse: NM_010153, AY686636, BC049279, BC029028, BC106091, and others.
Panther Classification:
Molecular Function -
transmembrane signal receptor
Gene Ontology Categories:
Function(s)
Process(es)
endocardial cushion development
negative regulation of cell adhesion
signal transduction
transmembrane receptor protein tyrosine kinase signaling pathway
epidermal growth factor receptor signaling pathway
axonogenesis
peripheral nervous system development
skeletal muscle tissue development
circadian rhythm
response to wounding
negative regulation of signal transduction
Schwann cell differentiation
phosphatidylinositol 3-kinase signaling
peptidyl-tyrosine phosphorylation
cranial nerve development
signal transduction by protein phosphorylation
cellular response to insulin stimulus
regulation of cell proliferation
odontogenesis
response to drug
negative regulation of neuron apoptotic process
tongue development
positive regulation of glucose import
negative regulation of secretion
neuron apoptotic process
mammary gland involution
positive regulation of protein tyrosine kinase activity
positive regulation of calcineurin-NFAT signaling cascade
extrinsic apoptotic signaling pathway in absence of ligand
protein tyrosine kinase activity
transmembrane receptor protein tyrosine kinase activity
receptor signaling protein tyrosine kinase activity
ATP binding
growth factor binding
protein tyrosine kinase activator activity
neuregulin receptor activity
neuregulin binding
identical protein binding
protein heterodimerization activity
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-green-659f68c6f4-2k58s:80/100.66.76.192:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0