Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ctla2b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001145801.1 | NP_001139273.1 | 2 | 208 | ||
NM_007797.1 | NP_031823.1 | 2 | 208 | |||
GenBank mRNA | AK150917.1 | - | 2 | 167 | ||
AY034578.1 | - | 2 | 148 | |||
X15592.1 | - | 2 | 148 |