Search
Search

Dlx6| Gene Symbol: Dlx6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_010057.2 | NP_034187.1 | 2 | 673 | ||
| GenBank mRNA | AB221684.1 | - | 2 | 261 | ||
| AF022078.1 | - | 2 | 210 | |||
| BC114342.1 | - | 2 | 257 | |||
| EF535989.1 | - | 2 | 673 | |||
| EF535990.1 | - | 3 | 528 | |||
| U67841.1 | - | 1 | 106 | |||